Learn More
HLA-G, Mouse, PE, Clone: 5A6G7, Abnova™
Mouse monoclonal antibody raised against synthetic peptide HLA-G.
Supplier: Abnova Corporation MAB4489
Description
HLA-G belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. HLA-G is expressed on fetal derived placental cells. The heavy chain is approximately 45kDa and its gene contains 8 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domain, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exon 6 encodes the cytoplasmic tail. [provided by RefSeq
Specifications
HLA-G | |
Monoclonal | |
0.1 mg/mL | |
Mouse monoclonal antibody raised against synthetic peptide HLA-G. | |
In PBS (0.2% BSA, 0.09% sodium azide) | |
MHC-G | |
Mouse | |
100 μg | |
Primary | |
This antibody allows to discriminate between soluble HLA-G protein yielded by shedding from membrane-bound HLA-G forms that do not contain the intron 4-encoded epitope and soluble HLA-G5/HLA-G6 produced from intron 4-retaining alternatively spliced mRNAs. HLA-G belongs to the MHC Class I molecules (MHC Class Ib; nonclassical) and it is expressed on the surface of trophoblast cells. | |
Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot | |
5A6G7 | |
PE | |
Flow Cytometry (10 ug/mL) The optimal working dilution should be determined by the end user. | |
HLA-G | |
HLA-G | |
A synthetic peptide (conjugated with OVA) corresponding to C-terminus of human soluble HLA-G5 and HLA-G6. | |
RUO | |
3135 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.