Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGB1, Mouse, Clone: J2E1, Abnova™

Catalog No. 89110615 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-615 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-615 Supplier Abnova Corporation Supplier No. MAB1771
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant HMGB1.

Specifications

Antigen HMGB1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone J2E1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant HMGB1.
Dilution Western Blot (1:500-1:2000) Immunohistochemistry (1:100-1:300) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene HMGB1
Gene Alias DKFZp686A04236/HMG1/HMG3/SBP-1
Gene Symbols HMGB1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-215 of human HMGB1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3146
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.