Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGB1, Mouse, Clone: PR45, Abnova™

Catalog No. 89113665 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-665 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-665 Supplier Abnova Corporation Supplier No. MAB8971
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of HMGB1.

Specifications

Antigen HMGB1
Applications Western Blot
Classification Monoclonal
Clone PR45
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of HMGB1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.02% sodium azide)
Gene HMGB1
Gene Alias DKFZp686A04236/HMG1/HMG3/SBP-1
Gene Symbols HMGB1
Host Species Mouse
Immunogen A synthetic peptide corresponding to HMGB1.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3146
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.