Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF4, Mouse, Clone: 3E17, Abnova™

Catalog No. 89106053 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-053 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-053 Supplier Abnova Corporation Supplier No. H00009324M05A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant HMGN3.

Thyroid hormone receptors are hormone-dependent transcription factors that regulate expression of a variety of specific target genes. The protein encoded by this gene binds thyroid hormone receptor beta, but only in the presence of thyroid hormone. The encoded protein, a member of the HMGN protein family, is thought to reduce the compactness of the chromatin fiber in nucleosomes, thereby enhancing transcription from chromatin templates. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MPKRKSPENTEGKDGSKVTKQEPTRRSARLSAKPAPPKPEPKPRKTSAKKEPGAKISRGAKGKKEEKQEAGKEGTEN

Specifications

Antigen KLF4
Applications ELISA
Classification Monoclonal
Clone 3E17
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant HMGN3.
Formulation ascites with no preservative
Gene HMGN3
Gene Accession No. BC009529
Gene Alias DKFZp686E20226/PNAS-24/PNAS-25/TRIP7
Gene Symbols HMGN3
Host Species Mouse
Immunogen HMGN3 (AAH09529,1 a.a. ∽ 77 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9324
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.