Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMOX1, Mouse, Clone: 1F12-A6, Abnova™

Catalog No. 89112961 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-961 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-961 Supplier Abnova Corporation Supplier No. MAB6628
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of HMOX1.

Heme oxygenase, an essential enzyme in heme catabolism, cleaves heme to form biliverdin, which is subsequently converted to bilirubin by biliverdin reductase, and carbon monoxide, a putative neurotransmitter. Heme oxygenase activity is induced by its substrate heme and by various nonheme substances. Heme oxygenase occurs as 2 isozymes, an inducible heme oxygenase-1 and a constitutive heme oxygenase-2. HMOX1 and HMOX2 belong to the heme oxygenase family. [provided by RefSeq

Specifications

Antigen HMOX1
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1F12-A6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of HMOX1.
Dilution Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene HMOX1
Gene Alias HO-1/HSP32/bK286B10
Gene Symbols HMOX1
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 1-30 of human HMOX1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3162
Test Specificity Detects a 32KDa protein, corresponding to the molecular mass of HO-1 on SDS Page immunoblots. Does not cross-react with HO-2
Target Species Bovine, Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.