Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hmox1, Mouse, Clone: GTS-1, Abnova™

Catalog No. 89110905 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-905 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-905 Supplier Abnova Corporation Supplier No. MAB2287
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Hmox1.

Specifications

Antigen Hmox1
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone GTS-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Hmox1.
Dilution ELISA (1:10000) Western Blot (5-10 ug/mL) Immunohistochemistry (Frozen sections) (5-10 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS, pH 7.4 (1% BSA, 0.09% sodium azide)
Gene Hmox1
Gene Alias HEOXG/Heox/Hmox/Ho-1/Ho1/hsp32
Gene Symbols Hmox1
Host Species Mouse
Immunogen Rat Hmox1 (heme oxygenase 1).
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24451
Test Specificity This antibody does not react with rabbit heme oxygenase I or heme oxygenase 2. This antibody will inhibit the activity of human or rat heme oxygenase 1.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water, store at -20°C with a recommended 0.1% sodium azide. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.