Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNRNPA2B1, Mouse, Clone: 1G12-6C5, Abnova™

Catalog No. 89104663 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-663 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-663 Supplier Abnova Corporation Supplier No. H00003181M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant HNRNPA2B1.

This gene belongs to the A/B subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has two repeats of quasi-RRM domains that bind to RNAs. This gene has been described to generate two alternatively spliced transcript variants which encode different isoforms. [provided by RefSeq

Sequence: MEREKEQFRKLFIGGLSFETTEESLRNYYEQWGKLTDCVVMRDPASKRSRGFGFVTFSSMAEVDAAMAARPHSIDGRVVEPKRAVAREESGKPGAHVTVKKLFVGGIKEDTEEHHLRDYFEEYGKIDTIEIITDRQSGKKRGFGFVTFDDHDPVDKIVLQKYHTINGHNAEVRKALSRQEMQEDLEVAILEVAPVMEEEEEDMVVEDLDMATRVGATEVVMTTMEEEIMEVEITMILEIITSNLLTTVQ

Specifications

Antigen HNRNPA2B1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1G12-6C5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant HNRNPA2B1.
Formulation ascites with no preservative
Gene HNRNPA2B1
Gene Accession No. BC000506
Gene Alias DKFZp779B0244/FLJ22720/HNRNPA2/HNRNPB1/HNRPA2/HNRPA2B1/HNRPB1/RNPA2/SNRPB1
Gene Symbols HNRNPA2B1
Host Species Mouse
Immunogen HNRNPA2B1 (AAH00506,1 a.a. ∽ 249 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3181
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.