Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNRNPA2B1, Mouse, Clone: DP3B3, Abnova™

Catalog No. 89111036 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-036 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-036 Supplier Abnova Corporation Supplier No. MAB2497
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant HNRNPA2B1.

This gene belongs to the A/B subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has two repeats of quasi-RRM domains that bind to RNAs. This gene has been described to generate two alternatively spliced transcript variants which encode different isoforms. [provided by RefSeq

Specifications

Antigen HNRNPA2B1
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone DP3B3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant HNRNPA2B1.
Dilution Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene HNRNPA2B1
Gene Alias DKFZp779B0244/FLJ22720/HNRNPA2/HNRNPB1/HNRPA2/HNRPA2B1/HNRPB1/RNPA2/SNRPB1
Gene Symbols HNRNPA2B1
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to human HNRNPA2B1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3181
Test Specificity Cross reacts with mouse for IHC on lung tissue. Reacts predominantly with a 36 KDa band (A2), & a 38 KDa band (B1).
Target Species Chicken, Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.