Learn More
HNRNPK, Mouse, Clone: F45 P9 C7, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of HNRNPK.
Supplier: Abnova Corporation MAB1891
Description
This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene is located in the nucleoplasm and has three repeats of KH domains that binds to RNAs. It is distinct among other hnRNP proteins in its binding preference; it binds tenaciously to poly(C). This protein is also thought to have a role during cell cycle progession. Several alternatively spliced transcript variants have been described for this gene, however, not all of them are fully characterized. [provided by RefSeq
Specifications
HNRNPK | |
Monoclonal | |
Unconjugated | |
Western Blot (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-5 ug/mL) The optimal working dilution should be determined by the end user. | |
HNRNPK | |
HNRNPK | |
A synthetic peptide corresponding to C-terminus of human HNRNPK. | |
RUO | |
3190 | |
Liquid |
Immunohistochemistry, Western Blot | |
F45 P9 C7 | |
Mouse monoclonal antibody raised against synthetic peptide of HNRNPK. | |
In PBS (0.08% sodium azide) | |
CSBP/FLJ41122/HNRPK/TUNP | |
Mouse | |
200 μg | |
Primary | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.