Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Homeobox protein CDX-2, Clone: 7D9.1,

Catalog No. MABE988MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE988MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE988MI Supplier MilliporeSigma™ Supplier No. MABE988
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Homeobox protein CDX-2 clone: 7D9.1 in Human samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

Homeobox protein CDX-2 (UniProt Q99626; also known as Caudal-type homeobox protein 2, Caudal type homeobox transcription factor 2, CDX-3) is encoded by the CDX2 (also known as CDX3, CDX2/AS) gene (Gene ID 1045) in human. CDX-2 is a transcription factor that plays an important role during embryonic development, where it determines intestinal identity in the presumptive gut endoderm. CDX-2 is also required for maintenance of intestinal identity in the intestinal epithelium throughout adulthood. Mitogen-activated protein kinases (MAPK) are reported to modulate CDX2 transcriptional activity and half-life via phosphorylation. Reduction of CDX2 expression facilitates tumor progression in murine models of colorectal cancer (CRC) and decreased CDX2 expression level in CRC correlates with poor prognosis in human.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Homeobox protein CDX-2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7D9.1
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q99626
Gene Symbols CDX2; CDX3
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human Homeobox protein CDX-2.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001256
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.