Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Horseradish peroxidase (HRP), Mouse, Clone: HP-03, Abnova™
Mouse monoclonal antibody raised against recombinant Horseradish peroxidase (HRP).
Supplier: Abnova Corporation MAB3616
Specifications
| Horseradish peroxidase (HRP) | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunocytochemistry, Western Blot | |
| HP-03 | |
| Mouse monoclonal antibody raised against recombinant Horseradish peroxidase (HRP). | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| Recombinant protein corresponding to Horseradish peroxidase (Grade I). | |
| RUO | |
| This antibody reacts with horseradish peroxidase. It is suitable for PAP detection system. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction