Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXA3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104664 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-664 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-664 Supplier Abnova Corporation Supplier No. H00003200A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant HOXA3.

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq

Sequence: MQKATYYDSSAIYGGYPYQAANGFAYNANQQPYPASAALGADGEYHRPACSLQSPSSAGGHPKAHELSEACLRTLSA

Specifications

Antigen HOXA3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant HOXA3.
Formulation 50% glycerol
Gene HOXA3
Gene Accession No. NM_153631
Gene Alias HOX1/HOX1E/MGC10155
Gene Symbols HOXA3
Host Species Mouse
Immunogen HOXA3 (NP_705895, 1 a.a. ∽ 77 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3200
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.