Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXA5, Mouse, Clone: 3G5, Abnova™

Catalog No. 89104666 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-666 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-666 Supplier Abnova Corporation Supplier No. H00003202M15A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant HOXA5.

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. Methylation of this gene may result in the loss of its expression and, since the encoded protein upregulates the tumor suppressor p53, this protein may play an important role in tumorigenesis. [provided by RefSeq

Sequence: TSTHSPQPDPLPCSAVAPSPGSDSHHGGKNSLSNSSGASADAGSTHISSREGVGTASGAEEDAPASSEQASAQSEPSPAPPAQPQIYPWMRKLHISHDNIGGPEGKRART

Specifications

Antigen HOXA5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3G5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant HOXA5.
Formulation ascites with no preservative
Gene HOXA5
Gene Accession No. NM_019102
Gene Alias HOX1/HOX1.3/HOX1C/MGC9376
Gene Symbols HOXA5
Host Species Mouse
Immunogen HOXA5 (NP_061975,91 a.a. ∽ 200 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3202
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.