Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXB8, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104671 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-671 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-671 Supplier Abnova Corporation Supplier No. H00003218A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant HOXB8.

This gene is a member of the Antp homeobox family and encodes a nuclear protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox B genes located on chromosome 17. The encoded protein functions as a sequence-specific transcription factor that is involved in development. Increased expression of this gene is associated with colorectal cancer. Mice that have had the murine ortholog of this gene knocked out exhibit an excessive pathologic grooming behavior. This behavior is similar to the behavior of humans suffering from the obsessive-compulsive spectrum disorder trichotillomania. [provided by RefSeq

Sequence: GESLRPNYYDCGFAQDLGGRPTVVYGPSSGGSFQHPSQIQEFYHGPSSLSTAPYQQNPCAVACHGDPGNFYGYDPLQRQSLFGAQDPDLVQYADCKLAA

Specifications

Antigen HOXB8
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant HOXB8.
Formulation 50% glycerol
Gene HOXB8
Gene Accession No. NM_024016
Gene Alias HOX2/HOX2D/Hox-2.4
Gene Symbols HOXB8
Host Species Mouse
Immunogen HOXB8 (NP_076921, 16 a.a. ∽ 114 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3218
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.