Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-HPRT, Clone: 2G8, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP174045100UL
Description
HPRT Monoclonal specifically detects HPRT in Human samples. It is validated for Western Blot, ELISA.Specifications
HPRT | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:5000, ELISA 1:100-1:2000 | |
NP_000185 | |
HPRT1 | |
Recombinant human HPRT (1-218aa) purified from E. coli | |
100 μL | |
Lipid and Metabolism | |
3251 | |
Human | |
Purified |
Western Blot, ELISA | |
2G8 | |
Unconjugated | |
PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide | |
EC 2.4.2.8, HGPRTHPRTHGPRTase, hypoxanthine phosphoribosyltransferase 1, hypoxanthine-guanine phosphoribosyltransferase | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human HPRT mAb, clone 2G8, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human HPRT protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction