Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRBL, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104682 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-682 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-682 Supplier Abnova Corporation Supplier No. H00003268A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant HRBL.

This gene is a member of the HIV-1 Rev binding protein (HRB) family and encodes a protein with one Arf-GAP zinc finger domain, several phe-gly (FG) motifs, and four asn-pro-phe (NPF) motifs. This protein interacts with Eps15 homology (EH) domains and plays a role in the Rev export pathway, which mediates the nucleocytoplasmic transfer of proteins and RNAs. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. The 3' UTR of this gene contains an insulin receptor substrate 3-like pseudogene. [provided by RefSeq]

Sequence: VKEFLQEKYEKKRWYVPPDQVKGPTYTKGSASTPVQGSIPEGKPLRTLLGDPAPSLSVAASTSSQPVSQSHARTSQARSTQPPPHSSVKKASTDLLAD

Specifications

Antigen HRBL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant HRBL.
Formulation 50% glycerol
Gene AGFG2
Gene Accession No. NM_006076
Gene Alias HRBL/RABR
Gene Symbols AGFG2
Host Species Mouse
Immunogen HRBL (NP_006067, 131 a.a. to 228 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3268
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.