Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HRP, Mouse, Clone: MAP A6-2, Abnova™

Catalog No. 89136037 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-037 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-037 Supplier Abnova Corporation Supplier No. MAB5673
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against HRP.

Specifications

Antigen HRP
Applications Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone MAP A6-2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against HRP.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS (50% glycerol)
Host Species Mouse
Immunogen Horse Radish Peroxidase.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water, store at -20°C for short term storage or at -80°C for long term storage. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.