Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD17B1, Mouse, Clone: 2E5, Abnova™

Catalog No. 89104687 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-687 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-687 Supplier Abnova Corporation Supplier No. H00003292M03A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant HSD17B1.

Sequence: VHTAFMEKVLGSPEEVLDRTDIHTFHRFYQYLAHSKQVFREAAQNPEEVAEVFLTALRAPKPTLRYFTTERFLPLLRMRLDDPSGSNYVTAMHREVF

Specifications

Antigen HSD17B1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2E5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant HSD17B1.
Formulation ascites with no preservative
Gene HSD17B1
Gene Accession No. NM_000413
Gene Alias EDH17B2/EDHB17/HSD17/MGC138140/SDR28C1
Gene Symbols HSD17B1
Host Species Mouse
Immunogen HSD17B1 (NP_000404,189 a.a. ∽ 285 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3292
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.