Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSF2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104688 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-688 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-688 Supplier Abnova Corporation Supplier No. H00003298A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant HSF2.

HSF2, as well as the related gene HSF1, encodes a protein that binds specifically to the heat-shock element and has homology to HSFs of other species. Heat shock transcription factors activate heat-shock response genes under conditions of heat or other stresses. Although the names HSF1 and HSF2 were chosen for historical reasons, these peptides should be referred to as heat-shock transcription factors. [provided by RefSeq

Sequence: MKQSSNVPAFLSKLWTLVEETHTNEFITWSQNGQSFLVLDEQRFAKEILPKYFKHNNMASFVRQLNMYGFRKVVHIDSGIVKQERDGPVEFQHPYFKQGQDDLLENIKRKVSSSKPEENKIRQEDLTKIISSAQKVQIKQETIESRLSELKSENESLWKEVSELRAKHAQQQQVIRKIVQFIVTLVQNNQLVSLKRKRPLLLNTNGAQKKNLFQHIVKEPTDNHHHKVIF

Specifications

Antigen HSF2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant HSF2.
Formulation 50% glycerol
Gene HSF2
Gene Accession No. BC005329
Gene Alias MGC117376/MGC156196/MGC75048
Gene Symbols HSF2
Host Species Mouse
Immunogen HSF2 (AAH05329, 1 a.a. ∽ 230 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3298
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.