Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP25, Mouse, Clone: HSP25-19, Abnova™

Catalog No. 89111174 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-174 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-174 Supplier Abnova Corporation Supplier No. MAB2968

Mouse monoclonal antibody raised against recombinant HSP25.

Specifications

Antigen HSP25
Applications ELISA, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone HSP25-19
Conjugate Unconjugated
Description heat shock protein 25
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene HSP25
Gene Symbols HSP25
Host Species Mouse
Immunogen Recombinant protein corresponding to chicken HSP25.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 428310
Target Species Chicken
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.