Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp90, Mouse, Clone: AC-16, Abnova™

Catalog No. 89135092 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-092 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-092 Supplier Abnova Corporation Supplier No. MAB2198
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Hsp90.

Specifications

Antigen Hsp90
Applications Western Blot
Classification Monoclonal
Clone AC-16
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Hsp90.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Host Species Mouse
Immunogen Native purified Hsp90 from water mold Achlya ambisexualis.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity It does not bind to the native form of Hsp90 and does not recognize E. coli and yeast Hsp90.
Target Species Avian, Chicken, Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.