Learn More
HSP90AA1, Mouse, Clone: 4F10, Abnova™
Mouse monoclonal antibody raised against full-length recombinant HSP90AA1.
Supplier: Abnova Corporation MAB1092
Description
HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. There are 2 major cytosolic HSP90 proteins, HSP90AA1, an inducible form, and HSP90AB1 (MIM 140572), a constitutive form. Other HSP90 proteins are found in endoplasmic reticulum (HSP90B1; MIM 191175) and mitochondria (TRAP1; MIM 606219) (Chen et al., 2005 [PubMed 16269234]).[supplied by OMIM
Specifications
HSP90AA1 | |
Monoclonal | |
Unconjugated | |
Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user. | |
HSP90AA1 | |
FLJ31884/HSP86/HSP89A/HSP90A/HSP90N/HSPC1/HSPCA/HSPCAL1/HSPCAL4/HSPN/Hsp89/Hsp90/LAP2 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
4F10 | |
Mouse monoclonal antibody raised against full length recombinant HSP90AA1. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
NM_005348 | |
HSP90AA1 | |
Recombinant protein corresponding to full length human HSP90AA1. | |
RUO | |
3320 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.