Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90AA1, Mouse, Clone: D7alpha, Abnova™

Catalog No. 89110884 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-884 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-884 Supplier Abnova Corporation Supplier No. MAB2197
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native HSP90AA1.

Specifications

Antigen HSP90AA1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone D7alpha
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native HSP90AA1.
Dilution Western Blot (1-5 ug/mL) Immunoprecipitation (5 ug) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene HSP90AA1
Gene Symbols HSP90AA1
Host Species Mouse
Immunogen Native purified HSP90AA1 from chicken brain.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 423463
Target Species Avian, Bovine, Chicken, Human, Mouse, Pig, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.