Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90AA1/HSP90AA2, Mouse, Clone: Hyb-K41220A, Abnova™

Catalog No. 89112963 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-963 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-963 Supplier Abnova Corporation Supplier No. MAB6631
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant HSP90AA1/HSP90AA2.

Specifications

Antigen HSP90AA1/HSP90AA2
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone Hyb-K41220A
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant HSP90AA1/HSP90AA2.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.09% sodium azide)
Gene Alias FLJ31884/HSP86/HSP89A/HSP90A/HSP90N/HSPC1/HSPCA/HSPCAL1/HSPCAL4/HSPN/Hsp89/Hsp90/LAP2
Gene Symbols HSP90AA1
Host Species Mouse
Immunogen Recombinant protein corresponding to human HSP90AA1/HSP90AA2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3320/3324
Test Specificity Detects 90kD proteins corresponding to the molecular mass of hsp90 alpha or beta.
Target Species Human, Rat, S. pombe, Yeast
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.