Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP90AA1 and HSP90AB1, Mouse, Clone: MBH90AB, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of HSP90AA1 and HSP90AB1.
Supplier: Abnova Corporation MAB6457
Description
Sequence: EEVHHGEEEVECSpecifications
HSP90AA1 and HSP90AB1 | |
Monoclonal | |
Unconjugated | |
Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μg | |
Primary | |
Bovine, Human, Mouse | |
Liquid |
Western Blot | |
MBH90AB | |
Mouse monoclonal antibody raised against synthetic peptide of HSP90AA1 and HSP90AB1. | |
In PBS (0.2% BSA, 0.09% sodium azide) | |
A synthetic peptide corresponding to amino acids at N-terminus of human HSP90AA1/HSP90AB1. | |
RUO | |
This antibody recognizes the epitope EEEVE within N-terminal part of ubiquitously expressed Hsp90a and Hsp90b proteins with calculated MW of 84.7 KDa and 83.3 KDa, respectively, however, migrating as 90 KDa bands under reducing SDS-PAGE conditions. | |
Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction