Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90AB1, Mouse, Clone: 5G4, Abnova™

Catalog No. 89108886 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-886 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-886 Supplier Abnova Corporation Supplier No. MAB10686
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant HSP90AB1.

HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. There are 2 major cytosolic HSP90 proteins, HSP90AA1 (MIM 140571), an inducible form, and HSP90AB1, a constitutive form. Other HSP90 proteins are found in endoplasmic reticulum (HSP90B1; MIM 191175) and mitochondria (TRAP1; MIM 606219) (Chen et al., 2005 [PubMed 16269234]).[supplied by OMIM

Specifications

Antigen HSP90AB1
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 5G4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant HSP90AB1.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene HSP90AB1
Gene Alias D6S182/FLJ26984/HSP90-BETA/HSP90B/HSPC2/HSPCB
Gene Symbols HSP90AB1
Host Species Mouse
Immunogen Recombinant protein corresponding to human HSP90AB1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3326
Target Species Human, Monkey, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.