Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90B1, Mouse, Clone: 2H3, Abnova™

Catalog No. 89109055 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-055 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-055 Supplier Abnova Corporation Supplier No. MAB1085
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant HSP90B1.

HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. HSP90B1 is an endoplasmic reticulum HSP90 protein. Other HSP90 proteins are found in cytosol (see HSP90AA1; MIM 140571) and mitochondria (TRAP1; MIM 606219) (Chen et al., 2005 [PubMed 16269234]).[supplied by OMIM

Specifications

Antigen HSP90B1
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 2H3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant HSP90B1.
Dilution Immunofluorescence (1:100-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene HSP90B1
Gene Accession No. NM_003299
Gene Alias ECGP/GP96/GRP94/TRA1
Gene Symbols HSP90B1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 676-803 of human HSP90B1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7184
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.