Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA1A, Mouse, ATTO390, Clone: 5A5, Abnova™

Catalog No. 89109553 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-553 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-553 Supplier Abnova Corporation Supplier No. MAB11391
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant HSPA1A.

This intronless gene encodes a 70kDa heat shock protein which is a member of the heat shock protein 70 family. In conjuction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. It is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which encode similar proteins. [provided by RefSeq

Specifications

Antigen HSPA1A
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5A5
Conjugate Atto 390
Description Mouse monoclonal antibody raised against recombinant HSPA1A.
Dilution Western Blot (1:1000) Immunofluorescence (1:500) Immunoprecipitation (1 ug) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene HSPA1A
Gene Alias FLJ54303/FLJ54370/FLJ54392/FLJ54408/FLJ75127/HSP70-1/HSP70-1A/HSP70I/HSP72/HSPA1/HSPA1B
Gene Symbols HSPA1A
Host Species Mouse
Immunogen Recombinant protein corresponding to human HSPA1A.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3303
Test Specificity Detects several members of the heat shock protein 70kDa gene family including Hsp70, Hsc70, Grp78 and following heat shock, Hsp72.
Target Species Amphibian, Chicken, Drosophila, Fish, Human, Mouse, Rat, Yeast
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.