Learn More
HSPA1A, Mouse, HRP, Clone: 5A5, Abnova™
Mouse monoclonal antibody raised against recombinant HSPA1A.
Supplier: Abnova Corporation MAB11403
Description
This intronless gene encodes a 70kDa heat shock protein which is a member of the heat shock protein 70 family. In conjuction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. It is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which encode similar proteins. [provided by RefSeq
Specifications
| HSPA1A | |
| Monoclonal | |
| HRP | |
| Western Blot (1:1000) Immunofluorescence (1:500) Immunoprecipitation (1 ug) The optimal working dilution should be determined by the end user. | |
| HSPA1A | |
| HSPA1A | |
| Recombinant protein corresponding to human HSPA1A. | |
| 100 μg | |
| Primary | |
| Detects several members of the heat shock protein 70kDa gene family including Hsp70, Hsc70, Grp78 and following heat shock, Hsp72. | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 5A5 | |
| Mouse monoclonal antibody raised against recombinant HSPA1A. | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| FLJ54303/FLJ54370/FLJ54392/FLJ54408/FLJ75127/HSP70-1/HSP70-1A/HSP70I/HSP72/HSPA1/HSPA1B | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 3303 | |
| Amphibian, Chicken, Drosophila, Fish, Human, Mouse, Rat, Yeast | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.