Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA2/HSPA8, Mouse, Clone: BB70, Abnova™

Catalog No. 89110883 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-883 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-883 Supplier Abnova Corporation Supplier No. MAB2196
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against HSPA2/HSPA8.

Specifications

Antigen HSPA2/HSPA8
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone BB70
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against HSPA2/HSPA8.
Dilution Western Blot (1 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene Symbols HSP70
Host Species Mouse
Immunogen Chicken HSPA2/HSP90 complex.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 423504/395853
Test Specificity This antibody detects 72 and 73 KDa proteins corresponding to the predicted molecular masses of HSPA2 and HSPA8, respectively. This antibody recognizes the inducible and constitutive forms of HSPA2 and does not cross-react with Hsp90.
Target Species Avian, Bovine, Canine, Chicken, Frog, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rat, Sheep, Yeast
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.