Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA4, Mouse, Clone: W27, Abnova™

Catalog No. 89111574 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-574 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-574 Supplier Abnova Corporation Supplier No. MAB3737
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native HSPA4.

Specifications

Antigen HSPA4
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone W27
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native HSPA4.
Dilution Immunoprecipitation (2 ug/mg protein lysate) Western Blot (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)(2-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide)
Gene HSPA4
Gene Alias APG-2/HS24/P52/MGC131852/RY/hsp70/hsp70RY
Gene Symbols HSPA4
Host Species Mouse
Immunogen Native purified HSPA4 from HeLa cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3308
Test Specificity This clone recognizes both the constitutive (Hsp73) and inducible (Hsp72) forms of Hsp70.
Target Species Bovine, Human, Monkey
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.