Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA4/HSPA8, Mouse, Clone: N27F3-4, Abnova™

Catalog No. 89112716 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-716 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-716 Supplier Abnova Corporation Supplier No. MAB6130
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant HSPA4/HSPA8.

Specifications

Antigen HSPA4/HSPA8
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone N27F3-4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant HSPA4/HSPA8.
Dilution Western Blot (1 ug/mL) Immunohistochemistry (1-10 ug/mL) Immunoprecipitation (1-10 ug/mL) Flow cytometry (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene Alias APG-2/HS24/P52/MGC131852/RY/hsp70/hsp70RY
Gene Symbols HSPA4
Host Species Mouse
Immunogen Recombinant protein corresponding to human HSPA4/HSPA8.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3308/3312
Test Specificity The epitope is in the region of amino acids 503-640 of human Hsp70.
Target Species Bovine, Canine, Chicken, Drosophila, Fish, Frog, Guinea Pig, Hamster, Human, Monkey, Mouse, Nematode, Plant, Pig, Rabbit, Rat, Sheep
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.