Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA8, Mouse, Clone: 21c3s, Abnova™

Catalog No. 89113599 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-599 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-599 Supplier Abnova Corporation Supplier No. MAB8819
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant HSPA8.

The product encoded by this gene belongs to the heat shock protein 70 family which contains both heat-inducible and constitutively expressed members. The latter are called heat-shock cognate proteins. This gene encodes a heat-shock cognate protein. This protein binds to nascent polypeptides to facilitate correct folding. It also functions as an ATPase in the disassembly of clathrin-coated vesicles during transport of membrane components through the cell. Two alternatively spliced variants have been characterized to date. [provided by RefSeq

Specifications

Antigen HSPA8
Applications ELISA
Classification Monoclonal
Clone 21c3s
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant HSPA8.
Dilution The optimal working dilution should be determined by the end user.
Formulation In citrate-Tris-HCl, pH 7.0 (0.02% Proclin 300)
Gene HSPA8
Gene Alias HSC54/HSC70/HSC71/HSP71/HSP73/HSPA10/LAP1/MGC131511/MGC29929/NIP71
Gene Symbols HSPA8
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human HSPA8.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3312
Test Specificity This antibody recognizes recombinant protein HSPA8.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.