Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPB8 (M04A), Mouse anti-Human, Clone: 5B12, Abnova™

Catalog No. 89106704 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-704 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-704 Supplier Abnova Corporation Supplier No. H00026353M04A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant HSPB8.

The protein encoded by this gene belongs to the superfamily of small heat-shock proteins containing a conservative alpha-crystallin domain at the C-terminal part of the molecule. The expression of this gene in induced by estrogen in estrogen receptor-positive breast cancer cells, and this protein also functions as a chaperone in association with Bag3, a stimulator of macroautophagy. Thus, this gene appears to be involved in regulation of cell proliferation, apoptosis, and carcinogenesis, and mutations in this gene have been associated with different neuromuscular diseases, including Charcot-Marie-Tooth disease. [provided by RefSeq

Sequence: KVCVNVHSFKPEELMVKTKDGYVEVSGKHEEKQQEGGIVSKNFTKKIQLPAEVDPVTVFASLSPEGLLIIEAPQVPPYSTFGESSFNNELPQDSQEVTCT

Specifications

Antigen HSPB8
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5B12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant HSPB8.
Formulation ascites with no preservative
Gene HSPB8
Gene Accession No. NM_014365
Gene Alias CMT2L/DHMN2/E2IG1/H11/HMN2/HMN2A/HSP22
Gene Symbols HSPB8
Host Species Mouse
Immunogen HSPB8 (NP_055180,97 a.a. ∽ 196 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26353
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.