Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPD1, Mouse, Clone: 1H8, Abnova™

Catalog No. 89111180 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-180 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-180 Supplier Abnova Corporation Supplier No. MAB2974

Mouse monoclonal antibody raised against recombinant HSPD1.

This gene encodes a member of the chaperonin family. The encoded mitochondrial protein may function as a signaling molecule in the innate immune system. This protein is essential for the folding and assembly of newly imported proteins in the mitochondria. This gene is adjacent to a related family member and the region between the 2 genes functions as a bidirectional promoter. Two pseudogenes, both located on chromosome 8, have been associated with this gene. Two transcript variants encoding the same protein have been identified for this gene. Mutations associated with this gene cause autosomal recessive spastic paraplegia 13. [provided by RefSeq

Specifications

Antigen HSPD1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1H8
Conjugate Unconjugated
Description heat shock 60kDa protein 1 (chaperonin)
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene HSPD1
Gene Alias CPN60, GROEL, HLD4, HSP60, HSP65, HuCHA60, SPG13
Gene Symbols HSPD1
Host Species Mouse
Immunogen Recombinant protein corresponding to human HSPD1.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3329
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.