Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPG2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104693 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-693 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-693 Supplier Abnova Corporation Supplier No. H00003339A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant HSPG2.

Heparan sulfate proteoglycan is a major component of basement membranes, where the molecule may be involved in the stabilization of other molecules as well as being involved with glomerular permeability to macromolecules and cell adhesion. This form of HSPG, known as HSPG2 or perlecan, is encoded by a gene that maps to chromosome 1. The gene for the form of HSPG associated with the cell surface of fibroblasts has been mapped to human chromosome 8 (MIM 142460).[supplied by OMIM

Sequence: GLRAYDGLSLPEDIETVTASQMRWTHSYLSDDEDMLADSISGDDLGSGDLGSGDFQMVYFRALVNFTRSIEYSPQLEDAGSREFREVSEAVVDTLESEYLKIPGDQVVSV

Specifications

Antigen HSPG2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant HSPG2.
Formulation 50% glycerol
Gene HSPG2
Gene Accession No. NM_005529
Gene Alias PLC/PRCAN/SJA/SJS/SJS1
Gene Symbols HSPG2
Host Species Mouse
Immunogen HSPG2 (NP_005520.3, 25 a.a. ∽ 134 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3339
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.