Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPH1, Mouse, Clone: J1G12, Abnova™

Catalog No. 89110777 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-777 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-777 Supplier Abnova Corporation Supplier No. MAB2035
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant HSPH1.

Specifications

Antigen HSPH1
Applications ELISA, Western Blot
Classification Monoclonal
Clone J1G12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant HSPH1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene HSPH1
Gene Alias DKFZp686M05240/HSP105/HSP105A/HSP105B/KIAA0201/NY-CO-25
Gene Symbols HSPH1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-858 of human HSPH1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10808
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.