Learn More
CD141 Mouse anti-Human, Brilliant Violet 650, Clone: 1A4, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 740604
Description
Reacts with a 75 kDa, a single chain, type I membrane glycoprotein referred to as thrombomodulin and fetomodulin. CD141 is expressed on endothelial cells, smooth muscle cells, epithelial cells, synovial lining cells, and keratinocytes. It is also found on megakaryocytes, peripheral blood moncytes, and neutrophils, but not on lymphocytes. Reports indicate that CD 141 plays an important role in Protein C activation and the initiation of the Protein C anticoagulant pathway. Thrombin loses its procagulant function when it binds to CD141, and the CD141/thrombin complex is able to activate Protein C.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.
Specifications
| CD141 | |
| Monoclonal | |
| 0.2mg/mL | |
| Thrombomodulin | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 1A4 | |
| Brilliant Violet 650 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only