Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD15 Mouse anti-Human, Brilliant Violet 650, Clone: W6D3, BD Optibuild™

Catalog No. BDB740613 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740613 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740613 Supplier BD Biosciences Supplier No. 740613
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The W6D3 monoclonal antibody specifically binds to 3-fucosyl-N-acetyllactosamine (3-FAL), a 220 kDa carbohydrate structure, also called X-hapten. 3-FAL is expressed on >95% of granulocytes, including neutrophils and eosinophils, and on monocytes to a varying degree, but not on lymphocytes or basophils. CD15 plays a role in mediating phagocytosis, bactericidal activity and chemotaxis. Most CD15 antibodies are IgM isotype; clone W6D3 is a mouse IgG1 isotype. In comparison studies with clone HI98, a known CD15 antibody, clone W6D3 shows brighter fluorescence staining and its binding can be blocked by clone HI98.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD15
Applications Flow Cytometry
Classification Monoclonal
Clone W6D3
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Description 3-fucosyl-N-acetyllactosamine; 3-FAL
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.