Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD21 Mouse anti-Human, Brilliant Violet 786, Clone: B-ly4, BD Optibuild™

Catalog No. BDB740969 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740969 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740969 Supplier BD Biosciences Supplier No. 740969
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The B-ly4 monoclonal antibody specifically binds to CD21, a 145 kDa glycosylated type I integral membrane protein. CD21 is a receptor for the C3d complement fragment and for Epstein-Barr virus (EBV). CD21 is expressed on mature B cells, follicular dendritic cells, and some epithelial cells. It is also weakly expressed on the subset of mature T cells and thymocytes. CD21 plays a role in B-cell activation and proliferation. It may also play a role in modulating the function of T cells in the immune response to infections by lymphotropic viruses. Recently, CD21 was found to be part of a large complex containing CD19, CD81, and possibly other molecules.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD21
Applications Flow Cytometry
Classification Monoclonal
Clone B-ly4
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Description CR2; Complement receptor type 2; C3DR; EBV-R; Epstein-Barr virus receptor
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.