Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD274 Mouse anti-Human, Brilliant Violet 510, Clone: MIH1, BD Optibuild™

Catalog No. BDB740190 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740190 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740190 Supplier BD Biosciences Supplier No. 740190
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PD-L1 and PD-L2 are newly discovered members of the B7 family are the ligands for the program death 1 (PD-1) receptor. They are expressed on immature dendritic cells and mature dendritic cells. PD-L1, also called B7-H1, is expressed on antigen-presenting cells, including IFN- γ-stimulated monocytes, and activated human and murine dendritic cells. Monoclonal antibodies that block PD-L1 and PD-L2 on dendritic cells result in enhanced T cell proliferation and cytokine production. PD-L1 is also expressed on placental trophoblasts, myocardial endothelium, cortical thymic epithelial cells, and on most carcinomas. Studies show overlapping functions of PD-L1 and PD-L2 and indicate an important role for the PD-L:PD-1 pathway in regulating T cell responses.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50-nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD274
Applications Flow Cytometry
Classification Monoclonal
Clone MIH1
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Description B7-H1, PD-L1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.