Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 Mouse anti-Human, BUV395, Clone: HIT3a, BD Optibuild™

Catalog No. BDB740283 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740283 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740283 Supplier BD Biosciences Supplier No. 740283
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HIT3a monoclonal antibody specifically binds to the human CD3 ε-chain, a 20 kDa subunit of the CD3/T cell antigen receptor complex found on 70-80% of normal human peripheral blood lymphocytes and 60-85% of thymocytes. Studies from the HLDA Workshop show that this antibody can be mitogenic for T lymphocytes. The CD3 complex plays a role in signal transduction during antigen recognition by the T cell receptor. HIT3a antibody does not stain intracellular CD3 unlike the other CD3-specific clone, UCHT1 (Cat. No. 555330/550368).

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348nm and an Em Max at 395nm, BD Horizon BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3
Applications Flow Cytometry
Classification Monoclonal
Clone HIT3a
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Description CD3E; T3E; TCRE; cd 3; cd-3; cd3; CD3-epsilon; 916
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.