Learn More
CD32 Mouse anti-Human, Brilliant Violet 605, Clone: FLI8.26 (2003), BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 740399
Description
The FLI8.26 monoclonal antibody specifically recognizes CD32 which is also known as Fcγreceptor type II (FcγRII). CD32 is a type I transmembrane glycoprotein that serves as a low affinity receptor for aggregated IgG. The FLI8.26 antibody recognizes a, b, and c forms of CD32 that are encoded by separate genes: CD32a/ FcγRIIa ( FCGR2A ), CD32b/ FcγRIIb ( FCGR2B ). These forms are differentially expressed on monocytes, granulocytes, T cells, B cells, NK cells, or platelets. These receptors play various roles in mediating and regulating inflammation and immunity including phagocytosis, cytotoxicity, degranulation, or platelet activation.
This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max of 602-nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20-nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605-nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser.

Specifications
CD32 | |
Monoclonal | |
0.2mg/mL | |
CD32a,FcγRIIa, FCGR2A; CD32b, FcγRIIb, FCGR2B; CD32c, FcγRIIc, FCGR2C | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG2b κ |
Flow Cytometry | |
FLI8.26 (2003) | |
Brilliant Violet 605 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
K562 or FcγRII+ L cell lines | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only