Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD33 Mouse anti-Human, BUV395, Clone: WM53 (also known as WM-53), BD Optibuild™

Catalog No. BDB740293 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740293 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740293 Supplier BD Biosciences Supplier No. 740293

Mouse Monoclonal Antibody

The WM53 monoclonal antibody specifically binds to human CD33. CD33 is a 67 kDa type I transmembrane glycoprotein. It is expressed on monocytes, activated T cells, myeloid progenitors, mast cells and weakly on polymorphonuclear cells. CD33 is absent on normal platelets, lymphocytes, erythrocytes, and hematopoietic stem cells. Reports indicate that this glycoprotein can function as a sialic acid-dependent cell adhesion molecule and this function can be modulated by endogenous sialoglycoconjugates when CD33 is expressed on the membrane.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348nm and an Em Max at 395nm, BD Horizon BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD33
Applications Flow Cytometry
Classification Monoclonal
Clone WM53 (also known as WM-53)
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Description Siglec-3; SIGLEC3; Sialic acid-binding Ig-like lectin 3; p67; gp67; My9
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Acute Myeloid Leukemia Blasts
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.