Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45RA Mouse anti-Human, Brilliant Violet 510, Clone: 5H9, BD Optibuild™

Catalog No. BDB740186 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740186 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740186 Supplier BD Biosciences Supplier No. 740186
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 5H9 monoclonal antibody specifically binds to the human form of the leukocyte common antigen (LCA), CD45RA. This clone also crossreacts with CD45RA expressed by peripheral blood lymphocytes, monocytes and some granulocytes from rhesus macaque, baboon, and cynomolgus monkeys. The observed frequency of CD45RA positive lymphocytes is higher in the non-human primate compared to what is seen with normal human donor lymphocytes.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50-nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45RA
Applications Flow Cytometry
Classification Monoclonal
Clone 5H9
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Description CD45R; PTPRC; LCA; Leukocyte common antigen
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.