Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD54 Mouse anti-Human, Brilliant Violet 605, Clone: HA58, BD Optibuild™

Catalog No. BDB740404 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740404 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740404 Supplier BD Biosciences Supplier No. 740404
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HA58 monoclonal antibody specifically binds to CD54 which is also known as ICAM-1 (intracellular adhesion molecule-1). CD54 is an 85-110 kDa type I transmembrane glycoprotein that belongs to the immunoglobulin supergene family. A soluble form of CD54 can also be found in biological fluids. CD54 is expressed on endothelial cells and both resting (weak) and activated (moderate) lymphocytes and monocytes. CD54 is a ligand for LFA-1 (CD11a/CD18) and Mac-1 (CD11b/CD18). The CD54 adhesion molecule plays roles in inflammatory and immune responses and neoplasia. This antibody (NA/LE format) blocks the mixed-lymphocyte reaction (MLR) and the purified format is suitable for staining acetone-fixed, frozen tissue sections.

This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max of 602-nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20-nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605-nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD54
Applications Flow Cytometry
Classification Monoclonal
Clone HA58
Concentration 0.2mg/mL
Conjugate Brilliant Violet 605
Description ICAM-1; intercellular adhesion molecule 1; BB2; P3.58
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen IFN-γ-treated BM314 colonic carcinoma cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.