Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD81 Mouse anti-Human, Brilliant Violet 711, Clone: JS-81, BD Optibuild™

Catalog No. BDB740789 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740789 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740789 Supplier BD Biosciences Supplier No. 740789
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The JS-81 monoclonal antibody specifically binds to CD81, which is also known as, Target of the antiproliferative antibody 1 (TAPA1, TAPA-1), or Tetraspanin-28 (Tspan-28/TSPAN28). CD81 is an ∽26 kDa transmembrane protein that belongs to the tetraspanin (TM4SF) family. It is involved in cell growth and signal transduction. CD81 has a very broad cellular distribution, being expressed on cells of hematopoietic, neuroectodermal and mesenchymal origin. In hematopoietic cells, the CD81 antigen is expressed on B and T lymphocytes, NK cells, thymocytes, eosinophils, germinal center follicular dendritic cells, and to a variable extent on monocytes. The CD81 antigen is not expressed on neutrophils, platelets, or erythrocytes. CD81-specific antibodies have been shown to have anti-proliferative effects on different lymphoid cell lines, particularly those derived from large cell lymphomas. They are also reported to induce homotypic cell aggregation. Immunoprecipitation studies reveal that CD81 is a component of a multimolecular complex of CD19, CD21, and CD225 that is involved in the activation and control of B cell growth.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD81
Applications Flow Cytometry
Classification Monoclonal
Clone JS-81
Concentration 0.2mg/mL
Conjugate Brilliant Violet 711
Description TAPA1; Tetraspanin-28; Tspan-28; TSPAN28; CVID6; S5.7; M38
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.