Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human, Clone: LN-3, MP Biomedicals™

Catalog No. MP08693031 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08693031 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP08693031 Supplier MP Biomedicals, Inc Supplier No. 08693031

Murine monoclonal antibody (IgG2b) reactive with a nonpolymorphic HLA-DR (Ia-like) antigen. The antigens show the typical 29-33 KDa molecular weight of Ia. The HLA-DR antigen is present on germinal center and mantle zone B cells, monocytes and macrophages and interdigitating histiocytes.

LN-3 is a murine monoclonal antibody (IgG2b) reactive with a nonpolymorphic HLA-DR (Ia-like) antigen. The antigens show the typical 29-33 K dalton molecular weight of Ia. The HLA-DR antigen is present on germinal center and mantle zone B cells, monocytes and macrophages and interdigitating histiocytes. T cells are generally negative although activated T cells may express HLA-DR.
TRUSTED_SUSTAINABILITY

Specifications

Applications Immunoassay
Classification Monoclonal
Clone LN-3
Conjugate Unconjugated
Description Murine monoclonal antibody (IgG2b) reactive with a nonpolymorphic HLA-DR (Ia-like) antigen. The antigens show the typical 29-33 KDa molecular weight of Ia. The HLA-DR antigen is present on germinal center and mantle zone B cells, monocytes and macrophages and interdigitating histiocytes.
Host Species Mouse
Immunogen HLA-DR
Quantity 2 mL
Regulatory Status RUO
Research Discipline Immunoassays
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerated
Form Liquid
Isotype IgG2b κ
Reactivity Human
Shipping Condition Cold Pack
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.