Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-A2 Mouse anti-Human, Brilliant Violet 510, Clone: BB7.2, BD Optibuild™

Catalog No. BDB740184 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740184 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740184 Supplier BD Biosciences Supplier No. 740184
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The monoclonal antibody BB7.2 specifically binds to the α subunit of the human leukocyte antigen-A2 (HLA-A2), a class I molecule of the major histocompatibility complex (MHC). The MHC gene locus encodes a group of highly polymorphic, cell-surface proteins that play a broad role in the immune response to protein antigens. MHC molecules bind and present small antigenic protein fragments to antigen-specific receptors expressed by T cells (TCR). Human ( human leukocyte antigen /HLA) MHC molecules are comprised of two major classes, MHC class I and class II. Functionally, class I MHC molecules bind peptides derived from intracellular antigens (eg, viral and some bacterial antigens) which are specifically recognized by CD8+ T cells. Class II MHC molecules bind antigens derived from pathogens multiplying in intracellular vesicles and ingested extracellular bacteria, both of which are recognized by CD4+ T cells. TCR recognize processed peptides bound to the MHC as well as regions of the MHC molecule itself. CD4 and CD8 accessory molecules strengthen the formation of the TCR-MHC complex through their interaction with non-polymorphic regions of the MHC molecule.

TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-A2
Applications Flow Cytometry
Classification Monoclonal
Clone BB7.2
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Description HLA class I histocompatibility antigen A2 alpha chain; HLA-A2
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.