Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG1 Fc Secondary Antibody (2C11), Novus Biologicals™
SDP

Catalog No. NB002824 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002824 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002824 Supplier Novus Biologicals Supplier No. NBP268313500UG

Mouse Monoclonal Antibody

IgG1 Fc Monoclonal specifically detects IgG1 Fc in Human samples. It is validated for ELISA.

Specifications

Antigen IgG1
Applications ELISA
Classification Monoclonal
Clone 2C11
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution ELISA
Formulation PBS
Host Species Mouse
Immunogen Purified human IgG
Quantity 500 μg
Test Specificity Mouse anti Human IgG1 antibody, clone 2C11, reacts with the gamma2-specific epitope inside the Fc fragment of human IgG1. It does not cross react with other human immunoglobulins.
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.